Anti-Malate Dehydrogenase Rabbit Polyclonal Antibody

Lieferant: US Biological
M2115-08G
USBIM2115-08GEA 809 EUR
USBIM2115-08G
Anti-Malate Dehydrogenase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Malate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR