Anti-Mannose 6 Phosphate Receptor, Cation-independent Mouse Monoclonal Antibody [clone: 8L641]

Lieferant: US Biological
M2255-16
USBIM2255-16EA 614 EUR
USBIM2255-16
Anti-Mannose 6 Phosphate Receptor, Cation-independent Mouse Monoclonal Antibody [clone: 8L641]
Antikörper
Type: Primary
Antigen: Mannose 6 Phosphate Receptor, Cation-independent
Clonality: monoclonal
Clone: 8L641
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR