Anti-Mannose 6 Phosphate Receptor, Cation-independent Mouse Monoclonal Antibody [clone: 10K74]

Lieferant: US Biological
M2255-17B
USBIM2255-17BEA 1060 EUR
USBIM2255-17B
Anti-Mannose 6 Phosphate Receptor, Cation-independent Mouse Monoclonal Antibody [clone: 10K74]
Antikörper
Recommended dilution: ELISA:2-8?g/ml

Type: Primary
Antigen: Mannose 6 Phosphate Receptor, Cation-independent
Clonality: monoclonal
Clone: 10K74
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR