Anti-MARCH2 Goat Polyclonal Antibody

Lieferant: US Biological
M2374-82A
USBIM2374-82AEA 460 EUR
USBIM2374-82A
Anti-MARCH2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000

Type: Primary
Antigen: MARCH2 (membrane-associated ring finger (C3HC4) 2, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR