Anti-MARCKS Rabbit Polyclonal Antibody

Lieferant: US Biological
M2375-03A
USBIM2375-03AEA 380 EUR
USBIM2375-03A
Anti-MARCKS Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: MARCKS (myristoylated alanine-rich protein kinase C substrate)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR