Anti-MARCKS Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
M2375-05J-BIOT
USBIM2375-05J-BIOTEA 884 EUR
USBIM2375-05J-BIOT
Anti-MARCKS Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: MARCKS (myristoylated alanine-rich protein kinase C substrate)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human