Anti-MARCKS Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
M2375-05J-PE
USBIM2375-05J-PEEA 884 EUR
USBIM2375-05J-PE
Anti-MARCKS Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: MARCKS (myristoylated alanine-rich protein kinase C substrate)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR