Anti-MARCO Mouse Monoclonal Antibody [clone: 9G228]

Lieferant: US Biological
M2378-01
USBIM2378-01EA 1010 EUR
USBIM2378-01
Anti-MARCO Mouse Monoclonal Antibody [clone: 9G228]
Antikörper
Recommended dilution: Immunofluorescence: 0. 6?g/ml

Type: Primary
Antigen: MARCO (macrophage receptor with collagenous structure)
Clonality: monoclonal
Clone: 9G228
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Bovine, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR