Anti-MARCO Mouse Monoclonal Antibody [clone: 10F59]
Lieferant: US Biological
M2378-02
USBIM2378-02EA 549 EUR
USBIM2378-02
Anti-MARCO Mouse Monoclonal Antibody [clone: 10F59]
Antikörper
Recommended dilution: Flow Cytometry:
Type: Primary
Antigen: MARCO (macrophage receptor with collagenous structure)
Clonality: monoclonal
Clone: 10F59
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Hamster