Anti-MARCO Rat Monoclonal Antibody [clone: 6A441]
Lieferant: US Biological
M2378-02E
USBIM2378-02EEA 681 EUR
USBIM2378-02E
Anti-MARCO Rat Monoclonal Antibody [clone: 6A441]
Antikörper
Recommended dilution: Flow Cytometry: 1:25-1:50; Immunochemistry: 1:200-1:400
Type: Primary
Antigen: MARCO (macrophage receptor with collagenous structure)
Clonality: monoclonal
Clone: 6A441
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse