Anti-MARCO Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A441]
Lieferant: US Biological
M2378-02F
USBIM2378-02FEA 561 EUR
USBIM2378-02F
Anti-MARCO Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A441]
Antikörper
Recommended dilution: Flow Cytometry: 1:2
Type: Primary
Antigen: MARCO (macrophage receptor with collagenous structure)
Clonality: monoclonal
Clone: 6A441
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse