Anti-MARCO Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6A441]

Lieferant: US Biological
M2378-02G
USBIM2378-02GEA 681 EUR
USBIM2378-02G
Anti-MARCO Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6A441]
Antikörper
Do not freeze!

Type: Primary
Antigen: MARCO (macrophage receptor with collagenous structure)
Clonality: monoclonal
Clone: 6A441
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR