Anti-MMP20 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
M2435-40-BIOT
USBIM2435-40-BIOTEA 884 EUR
USBIM2435-40-BIOT
Anti-MMP20 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: MMP20 (matrix metallopeptidase 20)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR