Anti-MBD2 Goat Polyclonal Antibody

Lieferant: US Biological
M2472-03B
USBIM2472-03BEA 601 EUR
USBIM2472-03B
Anti-MBD2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:64000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: MBD2 (methyl-CpG binding domain protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR