Anti-MCL1 Mouse Monoclonal Antibody [clone: 0.N.401]

Lieferant: US Biological
M2749-05-250
USBIM2749-05-250EA 361 EUR
USBIM2749-05-250
Anti-MCL1 Mouse Monoclonal Antibody [clone: 0.N.401]
Antikörper
Type: Primary
Antigen: MCL1 (myeloid cell leukemia 1)
Clonality: monoclonal
Clone: 0.N.401
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR