Anti-Melanoma Discriminating Antigen Mouse Monoclonal Antibody [clone: PAL-M1]

Lieferant: US Biological
M2869-40
USBIM2869-40EA 675 EUR
USBIM2869-40
Anti-Melanoma Discriminating Antigen Mouse Monoclonal Antibody [clone: PAL-M1]
Antikörper
Recommended dilution: Immunochemistry: 1:5-1:25

Type: Primary
Antigen: Melanoma Discriminating Antigen
Clonality: monoclonal
Clone: PAL-M1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR