Anti-Melatonin Rabbit Polyclonal Antibody

Lieferant: US Biological
M2871-04E
USBIM2871-04EEA 890 EUR
USBIM2871-04E
Anti-Melatonin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Melatonin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR