Anti-Melatonin Rat Polyclonal Antibody

Lieferant: US Biological
M2871-04F
USBIM2871-04FEA 795 EUR
USBIM2871-04F
Anti-Melatonin Rat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Melatonin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR