Anti-Meloxicam Sheep Polyclonal Antibody

Lieferant: US Biological
M2872-02B
USBIM2872-02BEA 1010 EUR
USBIM2872-02B
Anti-Meloxicam Sheep Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.156?g/ml

Type: Primary
Antigen: Meloxicam
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR