Anti-Membrane-type 1 Matrix Metalloproteinase Cytoplasmic Tail Binding Protein 1 Chicken Polyclonal Antibody
Lieferant: US Biological
M2873-36
USBIM2873-36EA 475 EUR
USBIM2873-36
Anti-Membrane-type 1 Matrix Metalloproteinase Cytoplasmic Tail Binding Protein 1 Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: Membrane-type 1 Matrix Metalloproteinase Cytoplasmic Tail Binding Protein 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human