Anti-Methotrexate Goat Polyclonal Antibody

Lieferant: US Biological
M3101-01A
USBIM3101-01AEA 381 EUR
USBIM3101-01A
Anti-Methotrexate Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Methotrexate
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR