Anti-MHC Class 1, H-2Dd, H-2Kv, H-2k/q/s Rat Monoclonal Antibody [clone: 10F87]

Lieferant: US Biological
M3885-63D
USBIM3885-63DEA 782 EUR
USBIM3885-63D
Anti-MHC Class 1, H-2Dd, H-2Kv, H-2k/q/s Rat Monoclonal Antibody [clone: 10F87]
Antikörper
Recommended dilution: Flow Cytometry: 1:100-1:200;

Type: Primary
Antigen: MHC Class 1, H-2Dd, H-2Kv, H-2k/q/s
Clonality: monoclonal
Clone: 10F87
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR