Anti-MHC Class 2 Mouse Monoclonal Antibody [clone: 10F99]
Lieferant: US Biological
M3887-09
USBIM3887-09EA 824 EUR
USBIM3887-09
Anti-MHC Class 2 Mouse Monoclonal Antibody [clone: 10F99]
Antikörper
Recommended dilution: Flow Cytometry:
Type: Primary
Antigen: MHC Class 2
Clonality: monoclonal
Clone: 10F99
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Cat