Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody [clone: 10F104]

Lieferant: US Biological
M3887-43C
USBIM3887-43CEA 549 EUR
USBIM3887-43C
Anti-MHC Class 2, DQ, DR, Polymorphic Mouse Monoclonal Antibody [clone: 10F104]
Antikörper
Recommended dilution: Flow Cytometry: 1:50-1:200;

Type: Primary
Antigen: MHC Class 2, DQ, DR, Polymorphic
Clonality: monoclonal
Clone: 10F104
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Goat, Human, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR