Anti-MHC Class 2, DR Mouse Monoclonal Antibody [clone: 10F111]

Lieferant: US Biological
M3887-46
USBIM3887-46EA 549 EUR
USBIM3887-46
Anti-MHC Class 2, DR Mouse Monoclonal Antibody [clone: 10F111]
Antikörper
Type: Primary
Antigen: MHC Class 2, DR
Clonality: monoclonal
Clone: 10F111
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR