Anti-MHC Class 2, I-Ak, d, b, q, r Rat Monoclonal Antibody [clone: 10F121]

Lieferant: US Biological
M3887-53
USBIM3887-53EA 771 EUR
USBIM3887-53
Anti-MHC Class 2, I-Ak, d, b, q, r Rat Monoclonal Antibody [clone: 10F121]
Antikörper
Recommended dilution: Flow Cytometry:

Type: Primary
Antigen: MHC Class 2, I-Ak, d, b, q, r
Clonality: monoclonal
Clone: 10F121
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR