Anti-MHC Class 2, I-Ak Mouse Monoclonal Antibody

Lieferant: US Biological
M3887-53E-500
USBIM3887-53E-500EA 546 EUR
USBIM3887-53E-500
Anti-MHC Class 2, I-Ak Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MHC Class 2, I-Ak
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR