Anti-MHC Class 2, I-Ak/s Mouse Monoclonal Antibody [clone: 10F125]

Lieferant: US Biological
M3887-54A
USBIM3887-54AEA 471 EUR
USBIM3887-54A
Anti-MHC Class 2, I-Ak/s Mouse Monoclonal Antibody [clone: 10F125]
Antikörper
Recommended dilution: Flow Cytometry: 1:50-100; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: MHC Class 2, I-Ak/s
Clonality: monoclonal
Clone: 10F125
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR