Anti-MHC Class 2, beta Chain DR Mouse Monoclonal Antibody [clone: 10F101]
Lieferant: US Biological
M3887-55R
USBIM3887-55REA 549 EUR
USBIM3887-55R
Anti-MHC Class 2, beta Chain DR Mouse Monoclonal Antibody [clone: 10F101]
Antikörper
Type: Primary
Antigen: MHC Class 2, beta Chain DR
Clonality: monoclonal
Clone: 10F101
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Sheep