Anti-Midkine Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
M3896-02-50
USBIM3896-02-50EA 425 EUR
USBIM3896-02-50
Anti-Midkine Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Midkine
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR