Anti-Mitocryptide-1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
M3990-75C1
USBIM3990-75C1EA 836 EUR
USBIM3990-75C1
Anti-Mitocryptide-1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Mitocryptide-1
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR