Anti-KMT2A Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
M4127-01A-BIOT
USBIM4127-01A-BIOTEA 884 EUR
USBIM4127-01A-BIOT
Anti-KMT2A Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: KMT2A (lysine (K)-specific methyltransferase 2A)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR