Anti-Monocarboxylate Transporter 3 Rabbit Polyclonal Antibody

Lieferant: US Biological
M4470-20-100
USBIM4470-20-100EA 742 EUR
USBIM4470-20-100
Anti-Monocarboxylate Transporter 3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Monocarboxylate Transporter 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR