Anti-Monocytes, Macrophages Mouse Monoclonal Antibody [clone: 5K102]

Lieferant: US Biological
M4520-15
USBIM4520-15EA 473 EUR
USBIM4520-15
Anti-Monocytes, Macrophages Mouse Monoclonal Antibody [clone: 5K102]
Antikörper
Type: Primary
Antigen: Monocytes, Macrophages
Clonality: monoclonal
Clone: 5K102
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR