Anti-Monocytes, Macrophages Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K102]
Lieferant: US Biological
M4520-17
USBIM4520-17EA 445 EUR
USBIM4520-17
Anti-Monocytes, Macrophages Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K102]
Antikörper
Do not freeze!
Type: Primary
Antigen: Monocytes, Macrophages
Clonality: monoclonal
Clone: 5K102
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken