Anti-Muscarinic Receptor M1 Rabbit Polyclonal Antibody

Lieferant: US Biological
M9520-03C
USBIM9520-03CEA 513 EUR
USBIM9520-03C
Anti-Muscarinic Receptor M1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Muscarinic Receptor M1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR