Anti-Myeloid Specific Antigen Mouse Monoclonal Antibody [clone: BM-5]

Lieferant: US Biological
M9759-60C
USBIM9759-60CEA 843 EUR
USBIM9759-60C
Anti-Myeloid Specific Antigen Mouse Monoclonal Antibody [clone: BM-5]
Antikörper
Type: Primary
Antigen: Myeloid Specific Antigen
Clonality: monoclonal
Clone: BM-5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR