Anti-Myokinase Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
M9835-10B
USBIM9835-10BEA 608 EUR
USBIM9835-10B
Anti-Myokinase Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Myokinase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen