Anti-Myosin Mouse Monoclonal Antibody [clone: 414]

Lieferant: US Biological
M9848-08
USBIM9848-08EA 624 EUR
USBIM9848-08
Anti-Myosin Mouse Monoclonal Antibody [clone: 414]
Antikörper
Recommended dilution: Immunochemistry: 1:5-1:10

Type: Primary
Antigen: Myosin
Clonality: monoclonal
Clone: 414
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR