Anti-Myosin Light Chain 2 Rabbit Polyclonal Antibody

Lieferant: US Biological
M9850-09C
USBIM9850-09CEA 425 EUR
USBIM9850-09C
Anti-Myosin Light Chain 2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: Myosin Light Chain 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR