Anti-N-Acyl Mannosamine-1-Dehydrogenase Goat Polyclonal Antibody

Lieferant: US Biological
N0008-01
USBIN0008-01EA 910 EUR
USBIN0008-01
Anti-N-Acyl Mannosamine-1-Dehydrogenase Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000-1:5000

Type: Primary
Antigen: N-Acyl Mannosamine-1-Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR