Anti-N-Acyl Mannosamine-1-Dehydrogenase Goat Polyclonal Antibody (Biotin)

Lieferant: US Biological
N0008-02
USBIN0008-02EA 744 EUR
USBIN0008-02
Anti-N-Acyl Mannosamine-1-Dehydrogenase Goat Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:7000-1:35000

Type: Primary
Antigen: N-Acyl Mannosamine-1-Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR