Anti-Nectin 3 Rat Monoclonal Antibody [clone: 103-A1]

Lieferant: US Biological
N0571-55D
USBIN0571-55DEA 944 EUR
USBIN0571-55D
Anti-Nectin 3 Rat Monoclonal Antibody [clone: 103-A1]
Antikörper
Recommended dilution: Flow Cytometry: 1:10; Immunochemistry: 1:10

Type: Primary
Antigen: Nectin 3
Clonality: monoclonal
Clone: 103-A1
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR