Anti-NERP-2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
N1073-55F
USBIN1073-55FEA 836 EUR
USBIN1073-55F
Anti-NERP-2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: NERP-2
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR