Anti-NERP-2 Rabbit Polyclonal Antibody (Rhodamine)

Lieferant: US Biological
N1073-55G
USBIN1073-55GEA 984 EUR
USBIN1073-55G
Anti-NERP-2 Rabbit Polyclonal Antibody (Rhodamine)
Antikörper
Type: Primary
Antigen: NERP-2
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR