Anti-Neomycin Phosphotransferase II Rabbit Polyclonal Antibody

Lieferant: US Biological
N2008-05B
USBIN2008-05BEA 830 EUR
USBIN2008-05B
Anti-Neomycin Phosphotransferase II Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: Neomycin Phosphotransferase II
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR