Anti-Neomycin Mouse Monoclonal Antibody [clone: 1.BB.912]

Lieferant: US Biological
N2009-100
USBIN2009-100EA 371 EUR
USBIN2009-100
Anti-Neomycin Mouse Monoclonal Antibody [clone: 1.BB.912]
Antikörper
Recommended dilution: ELISA: 1:1500

Type: Primary
Antigen: Neomycin
Clonality: Monoclonal
Clone: 1.BB.912
Conjugation: unconjugated
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR