Anti-Nesfatin-1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
N2058-53B
USBIN2058-53BEA 837 EUR
USBIN2058-53B
Anti-Nesfatin-1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Nesfatin-1
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR