Anti-NRG2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: US Biological
N2108-76-AP
USBIN2108-76-APEA 884 EUR
USBIN2108-76-AP
Anti-NRG2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: NRG2 (neuregulin 2)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human