Anti-Neurofilament, Subunit tail, Heavy Rabbit Polyclonal Antibody

Lieferant: US Biological
N2160-35-100
USBIN2160-35-100EA 526 EUR
USBIN2160-35-100
Anti-Neurofilament, Subunit tail, Heavy Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Neuro?lament, Subunit tail, Heavy
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR